Learn More
Raybiotech Inc Human PARC (CCL18) ELISA Kit
Supplier:  Raybiotech Inc ELHPARC1
Encompass_Preferred
For cell culture supernatants, plasma, and serum samples. Sensitivity: 2 pg/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: The antibody pair provided in this kit recognizes human PARC / CCL18.
Catalog No. 50-194-9620