Learn More
BIOHIPPO Human PARK7 Microsample
SDP
Catalog No. 5000315109
Encompass
Human PARK7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PARK7 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PARK7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PARK7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PARK7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15109 Supplier BIOHIPPO Supplier No. BHE15210879
Add to Cart
Add to Cart
Human PARK7 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PARK7 in serum plasma tissue homogenates cell lysates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 6 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PARK7 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PARK7 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PARK7 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color