Learn More
Description
PEP-159 is a 15 amino acid synthetic peptide whose sequence is from human parkin. The sequence of this peptide is (amino to carboxy terminus): C- I (298)-K-E-L-H-H-F-R-I-L- G-E-E-Q-Y-N (313) This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product and human parkin, catalog # PA1-751. Using a solution with equal weights per unit volume of peptide and corresponding antibody will yield a solution with a large molar excess of peptide that is able to competitively bind the antibody. Reconstitute with 0.1 mL of distilled water.
Specifications
Specifications
| Gene ID (Entrez) | 5071 |
| Species | Human |
| Purification Method | purified |
| Accession Number | O60260 |
| Common Name | Parkin |
| Concentration | 0.5 mg/mL |
| Content And Storage | -20°C. Avoid freeze-thaw cycles. |
| Description | Synthetic Peptide |
| Format | Lyophilized |
| Formulation | Peptide with no preservative |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.