Learn More
Biomatik Corporation Human Parkinson Disease Protein 7 (PARK7) High-sensitive ELISA Kit
Supplier:  Biomatik Corporation EKU10312
Encompass_Preferred
Human Parkinson Disease Protein 7 (PARK7) High-sensitive ELISA Kit for detecting PARK7 in serum, plasma, tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 15.6-1,000pg/ml. Sensitivity: 6.2pg/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: Q99497. Gene ID: 11315. Target Synonyms: DJ1; Autosomal Recessive,Early Onset. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-6636