Learn More
Sino Biological Anti-PARP-1 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 310193T46100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PARP / PARP1 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-240-8460