Learn More
Sino Biological Recombinant Human PARP-1 Protein (His & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 11040H49BB100
Encompass_Preferred
A DNA sequence encoding the Human PARP1 (NP_001609.2)(Met1-Trp1014) was expressed with a C-terminal polyhistidine tag followed by an AVI tag. The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-255-4958