Learn More
Sino Biological Anti-Parvin alpha/PARVA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13919T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Actopaxin (rh Actopaxin; Catalog#13919-H09E; Q9NVD7-1; Met1-Glu372). Actopaxin specific IgG was purified by Human Actopaxin affinity chromatography.
Catalog No. 50-255-4159