Learn More
Sino Biological Anti-Parvin alpha/PARVA Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 13919RP01100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Actopaxin (rh PARVA; Catalog#13919-H09E; Q9NVD7-1; Met1-Glu372). Total IgG was purified by Protein A affinity chromatography.
Catalog No. 50-255-4160