Learn More
Raybiotech Inc Human Pax-4 ELISA Kit
Supplier:  Raybiotech Inc ELHPAX41
Encompass_Preferred
For Serum, Plasma, and Cell Culture Supernatants. Sensitivity: 0.14 ng/ml. Pre-coated 96-well strip plate with additional reagents to allow for use in multiple experiments. Specificity: This ELISA antibody pair recognizes Human Pax-4.
Catalog No. 50-194-9622