Learn More
BIOHIPPO Human PAX6 Microsample
SDP
Catalog No. 5000313933
Encompass
Human PAX6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PAX6 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 098 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PAX6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PAX6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PAX6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13933 Supplier BIOHIPPO Supplier No. BHE15210694
Human PAX6 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PAX6 in Tissue homogenates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 098 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PAX6 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PAX6 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PAX6 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color