Learn More
Sino Biological Anti-PAX8 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 101554T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the N-terminus of the Human PAX8, and purified by antigen affinity chromatography.
Catalog No. 50-241-2388