Learn More
Biomatik Corporation Human Paxillin (PXN) ELISA Kit
Supplier:  Biomatik Corporation EKU06499
Encompass_Preferred
Human Paxillin (PXN) ELISA Kit for detecting PXN in tissue homogenates, cell lysates, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 0.156-10ng/ml. Sensitivity: 0.056ng/ml. Format: 96-Well Plate. Assay Type: Double-antibody Sandwich. Detection Method: Colormetric. Assay Time: 3h. Accession Number: P49023. Gene ID: 5829. Target Synonyms: Pax. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-5870