Learn More
Sino Biological Anti-PBK/TOPK Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10729MM10100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PBK / TOPK (rh PBK / TOPK; Catalog#10729-H08B; NP_060962.2; Met1-Val322). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7834