Learn More
Sino Biological Anti-PBK/TOPK Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10729RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PBK / TOPK (rh PBK; Catalog#10729-H08B; NP_060962.2; Met 1-Val 322). PBK specific IgG was purified by human PBK affinity chromatography.
Catalog No. 50-240-9817