Learn More
BIOHIPPO Human PC Microsample
SDP
Catalog No. 5000313964
Encompass
Human PC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PC in Serum Cell Lysate Hep Plasma EDTA Plasma Tissue Lysate It provides a detection range of 0 32-20 nmol/mL with a sensitivity of 0 122 nmol/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-13964 Supplier BIOHIPPO Supplier No. BHE15210554
Human PC Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PC in Serum Cell Lysate Hep Plasma EDTA Plasma Tissue Lysate It provides a detection range of 0 32-20 nmol/mL with a sensitivity of 0 122 nmol/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PC Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PC Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PC biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color