Learn More
Sino Biological Human PCDH18 PAb 100ul (Sino Biological)
Supplier:  Sino Biological 11640R077100
Encompass_Preferred
Produced in rabbits immunized with E. coli-derived Human PCDH18/Protocadherin 18 fragment, and purified by antigen affinity chromatography.
Catalog No. 50-237-4431