Learn More
BIOHIPPO Human PCII Microsample
SDP
Catalog No. 5000315842
Encompass
Human PCII Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCII in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PCII Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PCII Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PCII biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15842 Supplier BIOHIPPO Supplier No. BHE15211916
Human PCII Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCII in Serum plasma and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 115 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PCII Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PCII Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PCII biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color