Learn More
BIOHIPPO Human PCIII Microsample
SDP
Catalog No. 5000315325
Encompass
Human PCIII Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCIII in serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 65 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PCIII Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PCIII Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PCIII biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15325 Supplier BIOHIPPO Supplier No. BHE15211857
Human PCIII Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCIII in serum plasma tissue homogenates and other biological fluids It provides a detection range of 1 57-100 ng/mL with a sensitivity of 0 65 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PCIII Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PCIII Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PCIII biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color