Learn More
Abcam Human PCOLCE ELISA Kit, 1 x 96 Tests
Supplier:  Abcam AB3139211X96TESTS
Encompass_Preferred
Human PCOLCE ELISA Kit is a Sandwich (quantitative) ELISA kit for the measurement of Human PCOLCE in Human in Cell culture supernatant, Plasma, Serum samples.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. NC2480480