Learn More
BIOHIPPO Human PCP Microsample
SDP
Catalog No. 5000315211
Encompass
Human PCP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 03 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human P CP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human P CP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human P CP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15211 Supplier BIOHIPPO Supplier No. BHE15210697
Human PCP Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCP in serum plasma tissue homogenates and other biological fluids It provides a detection range of 6 25-400 ng/mL with a sensitivity of 2 03 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human P CP Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human P CP Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human P CP biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color