Learn More
Invitrogen™ Human PCPE-1 ELISA Kit
Description
Human PCPE-1 quantitates human PCPE-1 in serum, plasma, supernatant. The assay will exclusively recognize both natural and recombinant human PCPE-1.
Specifications
Specifications
| Accession Number | Q15113 |
| Assay Range | 51-12500 pg/mL |
| Assay Sensitivity | 51 pg/mL |
| Conjugate | HRP |
| Immunoassay Kit Format | Sandwich ELISA Kit |
| Product Type | ELISA |
| Sample Type | Plasma, Serum, Supernatant |
| Target Species | Human |
| For Use With (Equipment) | Absorbance Microplate Reader |
| Gene Alias | astroglial cell transcript 2; Astt2; Astt-2; P14; Pcolce; PCPE; PCPE1; PCPE-1; procollagen C-endopeptidase enhancer; procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen COOH-terminal proteinase enhancer 1; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; procollagen, type 1, COOH-terminal proteinase enhancer; type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancerastroglial cell transcript 2; Astt2; Astt-2; P14; Pcolce; PCPE; PCPE1; PCPE-1; procollagen C-endopeptidase enhancer; procollagen C-endopeptidase enhancer 1; procollagen C-endopeptidase enhancer protein; procollagen COOH-terminal proteinase enhancer 1; procollagen C-proteinase enhancer 1; procollagen C-proteinase enhancer protein; procollagen, type 1, COOH-terminal proteinase enhancer; type 1 procollagen C-proteinase enhancer protein; Type I procollagen COOH-terminal proteinase enhancer; |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.