Learn More
BIOHIPPO Human PCSK2 Microsample
SDP
Catalog No. 5000315425
Encompass
Human PCSK2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCSK2 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PCSK2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PCSK2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PCSK2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color
Catalog No. 50-003-15425 Supplier BIOHIPPO Supplier No. BHE15211461
Human PCSK2 Microsample ELISA Kit is a Sandwich-based immunoassay designed for the quantitative measurement of PCSK2 in tissue homogenates cell lysates and other biological fluids It provides a detection range of 0 32-20 ng/mL with a sensitivity of 0 113 ng/mL The assay can be completed in approximately 3 5h The procedure follows a The test principle applied in this kit is Sandwich enzyme immunoassay The microtiter plate provided in this kit has been pre-coated with an antibody specific to Human PCSK2 Standards or samples are added to the appropriate microtiter plate wells then with a biotin-conjugated antibody specific to Human PCSK2 Next Avidin conjugated to Horseradish Peroxidase (HRP) is added to each microplate well and incubated After TMB substrate solution is added only those wells that contain Human PCSK2 biotin-conjugated antibody and enzyme-conjugated Avidin will exhibit a change in color