Learn More
Sino Biological Human PCSK9 MAb 100ul (Sino Biological)
Supplier:  Sino Biological 10594MM20100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PCSK9 (rh PCSK9; Catalog#10594-H08H; AAV67948.1; Met1-Gln692). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-2302