Learn More
Sino Biological Anti-PCSK9 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10594MM20H100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PCSK9 (rh PCSK9; Catalog#10594-H08H; AAV67948.1; Met1-Gln692). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-241-2289