Learn More
Sino Biological Anti-PCSK9 Antibody, Mouse Monoclonal
Supplier:  Sino Biological 10594MM15100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PCSK9 (rh PCSK9; Catalog#10594-H08H; AAV67948.1; Met1-Gln692). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-240-7961