Learn More
Sino Biological Anti-PCSK9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 111006T08100
Encompass_Preferred
Produced in rabbits immunized with a synthetic peptide corresponding to the C terminus of the Human PCSK9, and purified by antigen affinity chromatography.
Catalog No. 50-240-8737