Learn More
Sino Biological Anti-PCSK9 Antibody, Rabbit Polyclonal
Supplier:  Sino Biological 10594T16100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PCSK9 / NARC1 (rh PCSK9 / NARC1; Catalog#10594-H08H; AAV67948.1; Met1-Gln692). PCSK9 / NARC1 specific IgG was purified by Human PCSK9 / NARC1 affinity chromatography.
Catalog No. 50-240-8296