Learn More
Sino Biological Recombinant Human PCSK9 Protein (D374Y, mFc Tag), Amine-Labeled Biotinylated
Supplier:  Sino Biological 29698H05H1B100
Encompass_Preferred
A DNA sequence encoding the human PCSK9 (NP_777596.2) (Met1-Gln692) was expressed with the Fc region of mouse IgG1 at the C-terminus. The purified protein was biotinylated in vitro.
Catalog No. 50-255-3824