Learn More
Carnabio Usa Inc Human PCTAIRE1, 10ug (2x5ug)
Catalog No. NC2607276
Encompass
Human PCTAIRE1, catalytic domain [107-496(end) amino acids and S153A of accession number NP_006192.1] was co-expressed as N-terminal GST-fusion protein (71 kDa) with GST-CyclinY(CCNY) [1-341(end) amino acids of accession number NP_659449.3] using baculovirus expression system. GSTPCTAIRE1 was purified by using glutathione sepharose chromatography
Catalog No. NC2607276 Supplier Carnabio Usa Inc Supplier No. 0411610
Human PCTAIRE1, catalytic domain [107-496(end) amino acids and S153A of accession number NP_006192.1] was co-expressed as N-terminal GST-fusion protein (71 kDa) with GST-CyclinY(CCNY) [1-341(end) amino acids of accession number NP_659449.3] using baculovirus expression system. GSTPCTAIRE1 was purified by using glutathione sepharose chromatography