Learn More
ACROBiosystems Human PD-1 / PDCD1 Full-Length Protein, C-terminal His Tag - 20 μg
Supplier:  ACROBiosystems PD1H52H620UG
Encompass_Preferred
Recombinant full-length human PD-1 (PDCD1) expressed in HEK293 cells and purified with a C-terminal polyhistidine tag. Supplied lyophilized in a stabilized formulation containing trehalose and detergent; verified for bioactivity by ELISA binding to PD-L1, PD-L2, and therapeutic anti-PD-1 antibodies. Suitable for assay development, ELISA, antibody characterization, and binding studies.
  • Full-length sequence expressed in HEK293 cells.
  • Lyophilized stabilized formulation containing trehalose and detergent.
  • Supplied as 20 μg; other sizes available.
  • Purity greater than 80% by SDS-PAGE.
  • Migrates ~40-120 kDa on SDS-PAGE due to glycosylation.
  • Endotoxin level <1.0 EU/μg.
  • Bioactivity confirmed by ELISA binding to PD-L1, PD-L2, and therapeutic anti-PD-1 antibodies.
  • Store lyophilized at -20°C or lower; after reconstitution store at -70°C and avoid repeated freeze-thaw cycles.
Catalog No. NC2575566