Learn More
R&D Systems™ Human PD-1 QuicKit ELISA

Specifications
Specifications
| Assay Range | 25.00 to 1600 pg/mL |
| Assay Sensitivity | 9.86 pg/mL |
| Gene ID (Entrez) | 5133 |
| Quantity | 1 Kit |
| Storage Requirements | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
| Synonym | CD279, CD279 antigen, hPD-1, PD1, PD1hPD-l, PDCD1, programmed cell death 1, programmed cell death protein 1, Protein PD-1, SLEB2 |
| Target | Human |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.