Learn More
ACROBiosystems Recombinant Protein;Biotinylated Human PD-1 / PDCD1 (recommended for biopanning), Avi Tag (Avitag™);HEK293;25UG;PD1,PDCD1,PD1,CD279,SLEB2,PDCD-1,PD-1,SLEB-2
Supplier:  ACROBiosystems PD1H82E425UG
Encompass_Preferred
Recombinant Protein;Biotinylated Human PD-1 / PDCD1 (recommended for biopanning), Avi Tag (Avitag™);25UG;Biotinylated Human PD-1, His Tag (PD1-H82E4) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accession # Q15116-1).;This protein carries an Avi tag (Avitag™) at the C-terminus, followed by a polyhistidine tag. The protein has a calculated MW of 18.6 kDa. The protein migrates as 35-45 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Nivolumab at 2 μg/mL (100 μL/well) can bind Biotinylated Human PD-1, His Tag (Cat. No. PD1-H82E4) with a linear range of 0.8-13 ng/mL (QC tested).
Catalog No. 50-108-9676