Learn More
ACROBiosystems ACROBiosystems Human PD-L1 / B7-H1 (19-134) Protein, His Tag (MALS verified)
Catalog No. 502107406
Encompass_Preferred
Recombinant Protein;1MG;Human PD-L1 (19-134), His Tag (PD1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Accession # Q9NZQ7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.2 kDa. The protein migrates as 17 kDa and 19 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1 (19-134), His Tag (Cat. No. PD1-H52H3) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human PD-L1 Antibody, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).
Catalog No. 50-210-7406 Supplier ACROBiosystems Supplier No. PD1H52H31MG
Recombinant Protein;1MG;Human PD-L1 (19-134), His Tag (PD1-H52H3) is expressed from human 293 cells (HEK293). It contains AA Phe 19 - Tyr 134 (Accession # Q9NZQ7-1).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 15.2 kDa. The protein migrates as 17 kDa and 19 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Immobilized Human PD-L1 (19-134), His Tag (Cat. No. PD1-H52H3) at 1 μg/mL (100 μL/well) can bind Monoclonal Anti-Human PD-L1 Antibody, Human IgG1 with a linear range of 0.1-3 ng/mL (QC tested).