Learn More
Medchemexpress LLC Human PD-L1 inhibitor I | 2135542-86-6 | 98.9% | 2350.58 | 10 MG
SDP
Catalog No. 5000423940
Encompass_Preferred
Human PD-L1 inhibitor I is a hPD-1 peptide ligand with a KD of 3.39 μM. It may disturb the binding of hPD-L1 to hPD-1.
  • hPD-1 peptide ligand
  • KD of 3.39 μM
  • May disturb the binding of hPD-L1 to hPD-1
Catalog No. 50-004-23940 Supplier Medchemexpress LLC Supplier No. HYP247410MG
Add to Cart
Add to Cart
Human PD-L1 inhibitor I is a hPD-1 peptide ligand with a KD of 3.39 μM. It may disturb the binding of hPD-L1 to hPD-1.
  • hPD-1 peptide ligand
  • KD of 3.39 μM
  • May disturb the binding of hPD-L1 to hPD-1