Learn More
Medchemexpress LLC Human PD-L1 inhibitor I | 2135542-86-6 | 98.9% | 2350.58 | 5 MG
SDP
Catalog No. 5000423646
Encompass_Preferred
The product is a peptide ligand that interacts with a protein involved in immune checkpoints. This peptide may disrupt the binding of two key proteins, with a dissociation constant (KD) of 3.39 μM. It is intended for research purposes only.
  • Biological activity: It functions as a ligand for the protein, demonstrating a binding affinity with a KD of 3.39 μM.
  • Mechanism of action: This peptide is thought to interfere with the interaction between two specific human proteins.
  • Physical characteristics: It appears as a solid, white to off-white in color.
  • Storage conditions: Sealed storage, protected from moisture and light, under nitrogen.
Catalog No. 50-004-23646 Supplier Medchemexpress LLC Supplier No. HYP24745MG
Add to Cart
Add to Cart
The product is a peptide ligand that interacts with a protein involved in immune checkpoints. This peptide may disrupt the binding of two key proteins, with a dissociation constant (KD) of 3.39 μM. It is intended for research purposes only.
  • Biological activity: It functions as a ligand for the protein, demonstrating a binding affinity with a KD of 3.39 μM.
  • Mechanism of action: This peptide is thought to interfere with the interaction between two specific human proteins.
  • Physical characteristics: It appears as a solid, white to off-white in color.
  • Storage conditions: Sealed storage, protected from moisture and light, under nitrogen.