Learn More
Medchemexpress LLC Human PD-L1 inhibitor II | 2135542-85-5 | 98.3% | 2219.45 | 10 MG
SDP
Catalog No. 5000423845
Encompass_Preferred
Human PD-L1 inhibitor II is a potent PD-L1 inhibitor with anti-cancer activity. It appears as a white to off-white solid.
  • Potent PD-L1 inhibitor with anti-cancer activity
  • White to off-white solid appearance
  • Soluble in H2O at 25 mg/mL (11.26 mM) with ultrasonic assistance
  • Ships at room temperature in continental US
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light, under nitrogen)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light, under nitrogen)
Catalog No. 50-004-23845 Supplier Medchemexpress LLC Supplier No. HYP247010MG
Add to Cart
Add to Cart
Human PD-L1 inhibitor II is a potent PD-L1 inhibitor with anti-cancer activity. It appears as a white to off-white solid.
  • Potent PD-L1 inhibitor with anti-cancer activity
  • White to off-white solid appearance
  • Soluble in H2O at 25 mg/mL (11.26 mM) with ultrasonic assistance
  • Ships at room temperature in continental US
  • Powder storage: -80°C for 2 years, -20°C for 1 year (sealed, away from moisture and light, under nitrogen)
  • In solvent storage: -80°C for 6 months, -20°C for 1 month (sealed, away from moisture and light, under nitrogen)