Learn More
Medchemexpress LLC Human PD-L1 inhibitor IV | 2135542-83-3 | 99.9% | 1856.90 | 10 MG
SDP
Catalog No. 5000423600
Encompass_Preferred
Human PD-L1 inhibitor IV is a polypeptide that acts as a competitive human PD-1 protein inhibitor with a Kd value of 1.38 μM. It inhibits the interaction of hPD-1/hPD-L1.
  • Acts as a competitive human PD-1 protein inhibitor
  • Inhibits the interaction of hPD-1/hPD-L1
  • Kd value of 1.38 μM for human PD-1 protein
Catalog No. 50-004-23600 Supplier Medchemexpress LLC Supplier No. HYP247710MG
Add to Cart
Add to Cart
Human PD-L1 inhibitor IV is a polypeptide that acts as a competitive human PD-1 protein inhibitor with a Kd value of 1.38 μM. It inhibits the interaction of hPD-1/hPD-L1.
  • Acts as a competitive human PD-1 protein inhibitor
  • Inhibits the interaction of hPD-1/hPD-L1
  • Kd value of 1.38 μM for human PD-1 protein