Learn More
Medchemexpress LLC Human PD-L1 inhibitor IV | 2135542-83-3 | 99.9% | 1856.90 | 5 MG
SDP
Catalog No. 5000424242
Encompass_Preferred
Human PD-L1 inhibitor IV is a polypeptide that acts as a competitive inhibitor of human PD-1 protein. It has a Kd value of 1.38 μM and is designed to inhibit the interaction between hPD-1 and hPD-L1. This product is for research use only.
  • Polypeptide composition
  • Competitive human PD-1 protein inhibitor
  • Kd value of 1.38 μM
  • Inhibits hPD-1/hPD-L1 interaction
  • For research use only
  • White to off-white solid
  • Requires sealed storage, away from moisture and light, under nitrogen
  • Soluble in DMSO at 50 mg/mL
Catalog No. 50-004-24242 Supplier Medchemexpress LLC Supplier No. HYP24775MG
Add to Cart
Add to Cart
Human PD-L1 inhibitor IV is a polypeptide that acts as a competitive inhibitor of human PD-1 protein. It has a Kd value of 1.38 μM and is designed to inhibit the interaction between hPD-1 and hPD-L1. This product is for research use only.
  • Polypeptide composition
  • Competitive human PD-1 protein inhibitor
  • Kd value of 1.38 μM
  • Inhibits hPD-1/hPD-L1 interaction
  • For research use only
  • White to off-white solid
  • Requires sealed storage, away from moisture and light, under nitrogen
  • Soluble in DMSO at 50 mg/mL