Learn More
Medchemexpress LLC Human PD-L1 inhibitor V | 2815311-61-4 | 98.7% | 1485.71 | 5 MG
SDP
Catalog No. 5000423846
Encompass_Preferred
Human PD-L1 inhibitor V is a human PD-1 protein binding peptide with a Kd value of 3.32 μM. It inhibits the interaction of hPD-1/hPD-L1. This product is for research use only and not for sale to patients.
Catalog No. 50-004-23846 Supplier Medchemexpress LLC Supplier No. HYP24785MG
Add to Cart
Add to Cart
Human PD-L1 inhibitor V is a human PD-1 protein binding peptide with a Kd value of 3.32 μM. It inhibits the interaction of hPD-1/hPD-L1. This product is for research use only and not for sale to patients.