Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Human PD-L1 inhibitor V | 2815311-61-4 | 98.70% | 1485.71 | 10 MG
SDP

Catalog No. 5000423939
Encompass_Preferred

Human PD-L1 inhibitor V is a human PD-1 protein binding peptide with a Kd value of 3.32 μM. It inhibits the interaction of hPD-1/hPD-L1. This peptide (Ar5M_2) is designed with anchor residues Y56, R113, A121, D122, and Y123. It is for research use only and not sold to patients.

  • Purity of 98.70%
  • Solid, white to off-white appearance
  • Sequence: Leu-Asp-Tyr-Val-Asn-Arg-Arg-Lys-Met-Tyr-Gln (LDYVNRRKMYQ)
  • Soluble in H2O
  • Kd value of 3.32 μM against human PD-1 protein

Catalog No. 50-004-23939 Supplier Medchemexpress LLC Supplier No. HYP247810MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Human PD-L1 inhibitor V is a human PD-1 protein binding peptide with a Kd value of 3.32 μM. It inhibits the interaction of hPD-1/hPD-L1. This peptide (Ar5M_2) is designed with anchor residues Y56, R113, A121, D122, and Y123. It is for research use only and not sold to patients.

  • Purity of 98.70%
  • Solid, white to off-white appearance
  • Sequence: Leu-Asp-Tyr-Val-Asn-Arg-Arg-Lys-Met-Tyr-Gln (LDYVNRRKMYQ)
  • Soluble in H2O
  • Kd value of 3.32 μM against human PD-1 protein

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.