Learn More
ACROBiosystems ACROBiosystems Human PD-L2 / B7-DC Protein, His Tag (SPR verified)
Supplier:  ACROBiosystems PD2H52201MG
Encompass_Preferred
Recombinant Protein;1MG;Human PD-L2, His Tag (SPR verified) (PD2-H5220) is expressed from human 293 cells (HEK293). It contains AA Leu 20 - Pro 219 (Accession # AAI13679).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 23.4 kDa. The protein migrates as 34-46 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.;Captured Human PD-L2, His Tag (SPR verified) (Cat. No. PD2-H5220) on CM5 Chip via anti-His antibody can bind Human PD-1, Fc Tag (Cat. No. PD1-H5257) with an affinity constant of 24.9 nM as determined in SPR assay (Biacore T200) (QC tested).
Catalog No. 50-210-6744