Learn More
Sino Biological Recombinant Human PD-1 Protein (ECD, hFc & AVI Tag), Biotinylated, HPLC-verified
Supplier:  Sino Biological 10377H41HB20
Encompass_Preferred
A DNA sequence encoding the human PDCD1 (NP_005009.2)(Met1-Gln167) was expressed with a c-terminal AVI tagged Fc region of human IgG1 at the C-terminus (Fc-AVI). The expressed protein was biotinylated in vivo by the Biotin-Protein ligase (BirA enzyme) which is co-expressed.
Catalog No. 50-224-6313