Learn More
Biomatik Corporation Human PD-1/PDCD1 (Programmed Cell Death Protein 1) ELISA Kit
Supplier:  Biomatik Corporation EKF58596
Encompass_Preferred
Human PD-1/PDCD1 (Programmed Cell Death Protein 1) ELISA Kit for detecting PD-1; PDCD1 in serum, plasma, cell culture supernatant, cell or tissue lysate, other biological fluids. Species Reactivity: Human (Homo sapiens). Detection Range: 31.25-2000pg/ml. Sensitivity: 18.75pg/ml. Format: 96-Well Plate. Assay Type: Sandwich ELISA, Double Antibody. Assay Time: 3h, 30 min. Accession Number: Q15116. Target Synonyms: PD-1; PDCD1; Programmed Cell Death Protein 1; CD279; PDCD1; CD279; programmed cell death 1; programmed cell death protein 1; Protein PD-1; SLEB2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-9168