Learn More
Sino Biological Recombinant Human PD-1 Protein (Site-Specific PE-Conjugated, ECD, His Tag)
Catalog No. 502992892
Encompass_Preferred
A DNA sequence encoding the extracellular domain (Met 1-Gln 167) of human PD1 (NP_005009.2) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with PE (Excitation = 495-566 nm, Emission Max.= 575 nm).
Catalog No. 50-299-2892 Supplier Sino Biological Supplier No. 10377H86HSP25
A DNA sequence encoding the extracellular domain (Met 1-Gln 167) of human PD1 (NP_005009.2) was expressed with a C-terminal polyhistidine tag. The protein is site-specifically conjugated with PE (Excitation = 495-566 nm, Emission Max.= 575 nm).