Learn More
Sino Biological Anti-PD-1 Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 10377MM18C100
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PD1 / PDCD1 (rh PD1 / PDCD1; Catalog#10377-H08H; NP_005009.2; Met1-Gln167) and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-7786