Learn More
Sino Biological Anti-PD-1 Antibody (PerCP), Mouse Monoclonal
Supplier:  Sino Biological 10377M140C100
Encompass_Preferred
This product is a recombinant monoclonal antibody expressed from HEK293 cells, and conjugated with PerCP under optimum conditions, the unreacted PerCP was removed.
Catalog No. 50-240-9956