Learn More
Sino Biological Anti-PD-1 Antibody (HRP), Mouse Monoclonal
Supplier:  Sino Biological 10377MM18H20
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PD1/PDCD1/CD279 (rh PD1/PDCD1/CD279; Catalog#10377-H08H; NP_005009.2; Met1-Gln167). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography and conjugated with horseradish-peroxidase (HRP).
Catalog No. 50-255-5346