Learn More
Sino Biological Anti-PD-1 Antibody (PE), Mouse Monoclonal
Supplier:  Sino Biological 10377MM18P25
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PD-1 (rh PD-1; Catalog#10377-H08H; NP_005009.2; Met1-Gln167) and conjugated with PE under optimum conditions, the unreacted PE was removed.
Catalog No. 50-241-2210