Learn More
Sino Biological Human PDCD1 MAb 20ul (Sino Biological)
Supplier:  Sino Biological 10377MM0320
Encompass_Preferred
This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human PD1 / PDCD1 / CD279 (rh PD1 / PDCD1 / CD279; Catalog#10377-H08H; NP_005009.2; Met1-Gln167). The IgG fraction of the cell culture supernatant was purified by Protein A affinity chromatography.
Catalog No. 50-237-1268