Learn More
Sino Biological Anti-PD-1 Antibody (Biotin), Rabbit Polyclonal
Supplier:  Sino Biological 10377RP03B100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human PD1 extracellular domain (rhPD1; Catalog#10377-H08H; Met 1-Gln 167; NP_005009.2). Total IgG was purified by Protein A affinity chromatography and then biotinylated.
Catalog No. 50-241-2197